Assay Details
| Gene Symbol: Bloc1s5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001107347.1 | NP_001100817.1 | 2 - 3 | 272 | 112 | |
| XM_006253807.2 | XP_006253869.1 | 2 - 3 | 417 | 112 | ||
| GenBank mRNA | BC166529.1 | - | 2 - 3 | 198 | 7189489 | 112 |
More Information
Updated Mapping Information
Set Membership: |
|

Bloc1s5
Genomic Map