Assay Details
| Gene Symbol: Slc35b3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003751664.3 | XP_003751712.1 | 5 - 6 | 782 | 79 | |
| XM_006253850.1 | XP_006253912.1 | 5 - 6 | 762 | 79 | ||
| XM_006253852.2 | XP_006253914.1 | 5 - 6 | 922 | 79 | ||
| XM_006253853.2 | XP_006253915.1 | 5 - 6 | 922 | 79 | ||
| XM_006253854.2 | XP_006253916.1 | 5 - 6 | 922 | 79 | ||
| XM_006253855.2 | XP_006253917.1 | 5 - 6 | 837 | 79 | ||
| XM_006253856.2 | XP_006253918.1 | 5 - 6 | 769 | 79 | ||
| XM_225253.8 | XP_225253.5 | 5 - 6 | 921 | 79 | ||
More Information
Set Membership: |
|

Slc35b3
Genomic Map