Assay Details
| Gene Symbol: Psmg4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001129879.2 | NP_001123351.1 | 1 - 2 | 271 | 79 | |
| XM_006253847.2 | XP_006253909.1 | 1 - 2 | 279 | 79 | ||
| GenBank mRNA | BC167096.1 | - | 1 - 2 | 198 | 8362489 | 79 |
| BE113476.1 | - | 2 - 3 | 79 | |||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Psmg4
Genomic Map