Assay Details
| Gene Symbol: LOC684107 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003751689.1 | XP_003751737.1 | 4 - 5 | 355 | 132 | |
| GenBank mRNA | FQ220106.1 | - | 1 - 2 | 104 | 132 | |
More Information
Set Membership: |
|

LOC684107
Genomic Map