Assay Details
| Gene Symbol: LOC684107 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003751689.1 | XP_003751737.1 | 5 - 6 | 540 | 153 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC684107| Gene Symbol: LOC684107 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003751689.1 | XP_003751737.1 | 5 - 6 | 540 | 153 | |
Set Membership: |
|