Assay Details
| Gene Symbol: Cdkal1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008771610.1 | XP_008769832.1 | 616 | 73 | ||
| XM_008771611.1 | XP_008769833.1 | 2261 | 73 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: Cdkal1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008771610.1 | XP_008769832.1 | 616 | 73 | ||
| XM_008771611.1 | XP_008769833.1 | 2261 | 73 | |||
Set Membership: |
|