Assay Details
| Gene Symbol: Gadd45g | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001077640.1 | NP_001071108.1 | 1 - 2 | 143 | 78 | |
| GenBank mRNA | AB020978.1 | - | 1 - 2 | 143 | 78 | |
| BC168643.1 | - | 1 - 2 | 143 | 9092341 | 78 | |
| FQ218856.1 | - | 1 - 2 | 161 | 78 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Gadd45g
Genomic Map