Assay Details
| Gene Symbol: LOC100911917 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003749430.3 | XP_003749478.1 | 490 | 83 | ||
| Gene Symbol: Med22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001077679.1 | NP_001071147.1 | 1 - 2 | 318 | 83 | |
| GenBank mRNA | BC127519.1 | - | 1 - 2 | 318 | 7104131 | 83 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

LOC100911917
Genomic Map