Assay Details
| Gene Symbol: LOC100911917 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003749430.3 | XP_003749478.1 | 571 | 79 | ||
| Gene Symbol: Med22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001077679.1 | NP_001071147.1 | 2 - 3 | 399 | 79 | |
| GenBank mRNA | BC127519.1 | - | 2 - 3 | 399 | 7104131 | 79 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

LOC100911917
Genomic Map