Assay Details
| Gene Symbol: LOC102554393 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006233879.2 | XP_006233941.2 | 28 - 29 | 4150 | 78 | |
| Gene Symbol: Cacfd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001106561.1 | NP_001100031.1 | 2 - 3 | 269 | 78 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001106561.1 transcripts.
Set Membership: |
|

LOC102554393
Genomic Map