Assay Details
| Gene Symbol: Ptges2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001107832.1 | NP_001101302.1 | 5 - 6 | 1395 | 79 | |
| GenBank mRNA | FQ231959.1 | - | 5 - 6 | 890 | 79 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ptges2
Genomic Map