Assay Details
| Gene Symbol: Ptges2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001107832.1 | NP_001101302.1 | 1 - 2 | 790 | 142 | |
| GenBank mRNA | FQ231959.1 | - | 1 - 2 | 283 | 142 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Ptges2
Genomic Map