Assay Details
| Gene Symbol: Brd3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001108575.1 | NP_001102045.1 | 9 - 10 | 1838 | 55 | |
| XM_006233825.2 | XP_006233887.1 | 10 - 11 | 1892 | 55 | ||
| XM_006233826.2 | XP_006233888.1 | 10 - 11 | 1918 | 55 | ||
| XM_006233830.2 | XP_006233892.1 | 9 - 10 | 1841 | 55 | ||
| GenBank mRNA | BC168949.1 | - | 10 - 11 | 1837 | 7112751 | 55 |
More Information
Updated Mapping Information
Set Membership: |
|

Brd3
Genomic Map