Assay Details
| Gene Symbol: Rabl6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001108573.1 | NP_001102043.1 | 10 - 11 | 1533 | 91 | |
| XM_008761603.1 | XP_008759825.1 | 10 - 11 | 1229 | 91 | ||
| XM_008761604.1 | XP_008759826.1 | 7 - 8 | 992 | 91 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Rabl6
Genomic Map