Assay Details
| Gene Symbol: Sh3glb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001009692.1 | NP_001009692.1 | 4 - 5 | 572 | 72 | |
| XM_006233816.2 | XP_006233878.1 | 4 - 5 | 549 | 72 | ||
| XM_006233817.2 | XP_006233879.1 | 4 - 5 | 549 | 72 | ||
| XM_006233818.2 | XP_006233880.1 | 4 - 5 | 548 | 72 | ||
| XM_006233819.2 | XP_006233881.1 | 4 - 5 | 548 | 72 | ||
| XM_006233820.2 | XP_006233882.1 | 4 - 5 | 548 | 72 | ||
| XM_006233822.2 | XP_006233884.1 | 4 - 5 | 548 | 72 | ||
| XM_008761661.1 | XP_008759883.1 | 4 - 5 | 549 | 72 | ||
| XM_008761662.1 | XP_008759884.1 | 4 - 5 | 548 | 72 | ||
| GenBank mRNA | BC087652.1 | - | 4 - 5 | 572 | 7312960 | 72 |
More Information
Updated Mapping Information
Set Membership: |
|

Sh3glb2
Genomic Map