Assay Details
| Gene Symbol: Pbx3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001107834.1 | NP_001101304.1 | 8 - 9 | 1262 | 135 | |
| XM_006233975.2 | XP_006234037.1 | 7 - 8 | 1211 | 135 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
regulation of respiratory gaseous exchange by neurological system process
regulation of transcription, DNA-templated
transcription from RNA polymerase II promoter
respiratory gaseous exchange
adult locomotory behavior
dorsal spinal cord development
positive regulation of transcription from RNA polymerase II promoter
neuron development
RNA polymerase II distal enhancer sequence-specific DNA binding
transcriptional activator activity, RNA polymerase II distal enhancer sequence-specific binding
transcription factor activity, sequence-specific DNA binding
sequence-specific DNA binding
regulation of respiratory gaseous exchange by neurological system process
regulation of transcription, DNA-templated
transcription from RNA polymerase II promoter
respiratory gaseous exchange
adult locomotory behavior
dorsal spinal cord development
positive regulation of transcription from RNA polymerase II promoter
neuron development
RNA polymerase II distal enhancer sequence-specific DNA binding
transcriptional activator activity, RNA polymerase II distal enhancer sequence-specific binding
transcription factor activity, sequence-specific DNA binding
sequence-specific DNA binding
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P532754 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 152256 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 355988 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 404465 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 675911 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 226518 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 326648 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 724525 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 227892 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 298283 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 255196 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 860679 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 737546 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 518550 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 685662 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 137207 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 290189 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 786576 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 183562 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 952610 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 128466 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 536143 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 310705 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 332284 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 235140 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 136168 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 936085 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 814367 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 144225 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 829232 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 624831 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 264820 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 927584 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 610462 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 736059 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 671393 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 675925 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 343151 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 825018 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 625214 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 189638 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 935810 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 880956 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 328291 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 728638 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 639987 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 915656 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 791257 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 27275 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 623375 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 893355 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 395011 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 550632 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 136341 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 755740 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 503574 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 802934 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 121985 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 948443 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 482162 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 589935 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 797573 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 771897 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 637312 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 159709 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 396141 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 255775 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 635914 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 649786 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 651367 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 619 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 550467 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 266892 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 456038 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 241422 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 71522 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 916859 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 519905 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 127190 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 263146 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 50630 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 603882 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 158169 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 876164 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 822128 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 170418 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 776344 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 17527 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 738983 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 192711 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 118980 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 565657 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 6966 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 46783 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 323243 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 665777 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 648280 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 276648 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 316529 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 556720 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 81906 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 47550 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 357376 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 638400 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 1021 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 168513 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 334954 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 665660 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 805970 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 797793 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 285344 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 558498 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 552108 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 233201 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 972022 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 402354 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 200525 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 419190 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 758899 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 687759 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 95988 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 130214 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 34350 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 78936 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 57763 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 19781 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 120948 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 119014 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 62982 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 49380 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 47617 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 38976 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 151592 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 96805 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 100354 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 44276 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 7779 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 150588 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 68845 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 25427 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 150661 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 60622 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 21066 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 123273 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 85394 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P532754 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P532754 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P532754 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_42_P475458 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P161622 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P532754 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 72002 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 83235 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 25417 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 13517 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 22556 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 47321 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10844536 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10844533 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10844535 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10844537 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10844532 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10844538 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10844539 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10844534 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10844531 | - | - |
| Rat Genome 230 2.0 Array | 1374503_at | - | - |
| Rat Genome 230 2.0 Array | 1374503_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1374503_at | - | - |
| Affymetrix HT Rat Focus Array | 1374503_PM_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR0300022554.rn.1 | TC0300002912.rn.1 | RTA TC |
| Rat Expression Set 230 | 1374503_at | - | - |
| Rat Expression Set 230 | 1374503_PM_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10844531 | - | - |
| Rat Genome U34 Set | rc_AI029675_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 22029382 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1357931 | - | - |

Pbx3
Genomic Map