Assay Details
| Gene Symbol: Abl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001100850.1 | NP_001094320.1 | 4 - 5 | 2137 | 90 | |
| XM_006233918.2 | XP_006233980.1 | 4 - 5 | 934 | 90 | ||
| GenBank mRNA | DQ294401.1 | - | 4 - 5 | 879 | 90 | |
| DQ294402.1 | - | 4 - 5 | 879 | 90 | ||
| Gene Symbol: LOC100909750 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003749439.3 | XP_003749487.1 | 934 | 90 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

Abl1
Genomic Map