Assay Details
| Gene Symbol: Uck1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001107831.3 | NP_001101301.2 | 4 - 5 | 596 | 62 | |
| GenBank mRNA | BC168897.1 | - | 4 - 5 | 535 | 8362491 | 62 |
More Information
Updated Mapping Information
Set Membership: |
|

Uck1
Genomic Map