Assay Details
| Gene Symbol: Entpd8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033565.1 | NP_001028737.1 | 1 - 2 | 131 | 112 | |
| XM_006233639.2 | XP_006233701.1 | 2 - 3 | 388 | 112 | ||
| XM_006233640.2 | XP_006233702.1 | 3 - 4 | 482 | 112 | ||
| XM_006233642.2 | XP_006233704.1 | 2 - 3 | 388 | 112 | ||
| XM_006233643.2 | XP_006233705.1 | 2 - 3 | 388 | 112 | ||
| GenBank mRNA | AY536920.1 | - | 1 - 2 | 131 | 112 | |
More Information
Updated Mapping Information
Set Membership: |
|

Entpd8
Genomic Map