Assay Details
| Gene Symbol: RGD1311084 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006233914.2 | XP_006233976.1 | 6 - 7 | 1232 | 72 | |
| GenBank mRNA | FQ214464.1 | - | 5 - 6 | 1003 | 72 | |
More Information
Set Membership: |
|

RGD1311084
Genomic Map