Assay Details
| Gene Symbol: Clic3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001013080.2 | NP_001013098.2 | 4 - 5 | 388 | 93 | |
| XM_006233590.2 | XP_006233652.1 | 4 - 5 | 370 | 93 | ||
| XM_008761584.1 | XP_008759806.1 | 3 - 4 | 1076 | 93 | ||
| XM_008761585.1 | XP_008759807.1 | 4 - 5 | 1031 | 93 | ||
| GenBank mRNA | CF115110.1 | - | 4 - 5 | 388 | 93 | |
| CK841958.1 | - | 2 - 3 | 93 | |||
More Information
Updated Mapping Information
Set Membership: |
|

Clic3
Genomic Map