Assay Details
| Gene Symbol: Triqk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001171802.1 | NP_001165273.1 | 1 - 2 | 116 | 128 | |
| XM_006237906.2 | XP_006237968.1 | 1 - 2 | 130 | 128 | ||
| GenBank mRNA | BC089993.1 | - | 1 - 2 | 86 | 7319652 | 128 |
| CB805415.1 | - | 1 - 2 | 116 | 128 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the BC089993.1, NM_001171802.1, XM_006237906.2 transcripts.
Set Membership: |
|

Triqk
Genomic Map