Assay Details
| Gene Symbol: Gja10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001173508.1 | NP_001166979.1 | 1 | 1462 | 75 | |
| XM_008763619.1 | XP_008761841.1 | 2 | 1841 | 75 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Gja10
Genomic Map