Assay Details
| Gene Symbol: Hint2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001107955.2 | NP_001101425.2 | 1 - 2 | 80 | 76 | |
| GenBank mRNA | BI282127.1 | - | 4 - 5 | 76 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Hint2
Genomic Map