Assay Details
| Gene Symbol: LOC100910771 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003750753.3 | XP_003750801.1 | 10 - 11 | 1214 | 103 | |
| XM_006245614.2 | XP_006245676.1 | 10 - 11 | 1214 | 103 | ||
| Gene Symbol: Map3k7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001107920.2 | NP_001101390.2 | 10 - 11 | 1238 | 103 | |
| XM_006237966.2 | XP_006238028.1 | 10 - 11 | 2430 | 103 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

LOC100910771
Genomic Map