Assay Details
| Gene Symbol: Arhgef39 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001106676.2 | NP_001100146.2 | 8 - 9 | 1007 | 61 | |
| GenBank mRNA | BC158673.1 | - | 8 - 9 | 1007 | 8371589 | 61 |
More Information
Updated Mapping Information
Set Membership: |
|

Arhgef39
Genomic Map