Assay Details
| Gene Symbol: Ccl27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001108660.1 | NP_001102130.1 | 2 - 3 | 420 | 142 | |
| XM_006238073.2 | XP_006238135.1 | 5 - 6 | 373 | 142 | ||
| XM_006238074.2 | XP_006238136.1 | 5 - 6 | 370 | 142 | ||
| XM_006238076.1 | XP_006238138.1 | 3 - 4 | 199 | 142 | ||
| XM_006238077.2 | XP_006238139.1 | 3 - 4 | 212 | 142 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ccl27
Genomic Map