Assay Details
| Gene Symbol: Ccl27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006238079.2 | XP_006238141.1 | 3 - 4 | 214 | 122 | |
| XM_008763627.1 | XP_008761849.1 | 5 - 6 | 374 | 122 | ||
More Information
Set Membership: |
|

Ccl27
Genomic Map