Assay Details
| Gene Symbol: RGD1305420 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001025652.1 | NP_001020823.1 | 1 - 2 | 129 | 94 | |
| GenBank mRNA | BC097981.1 | - | 1 - 2 | 129 | 7443366 | 94 |
More Information
Updated Mapping Information
Set Membership: |
|

RGD1305420
Genomic Map