Assay Details
| Gene Symbol: Rragd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001106641.2 | NP_001100111.1 | 6 - 7 | 1142 | 80 | |
| GenBank mRNA | BC167014.1 | - | 6 - 7 | 1142 | 9034554 | 80 |
More Information
Updated Mapping Information
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P1041460 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 853853 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 950149 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 747502 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 449198 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 851133 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 627265 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 912911 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 90278 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 902386 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 734798 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 806347 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 756501 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 472736 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 10384 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 231762 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 170945 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 857586 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 561546 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 961161 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 568388 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 30836 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 160284 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 279008 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 494355 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 120475 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 326302 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 458664 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 624 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 107155 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 167641 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 57776 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 36959 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 26505 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P1041460 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P1041460 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P1041460 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P1041460 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 91032 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10868133 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10868129 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10868128 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10868130 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10868131 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10868132 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10868125 | - | - |
| Rat Genome 230 2.0 Array | 1373427_at | - | - |
| Rat Genome 230 2.0 Array | 1373427_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1373427_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1373427_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR0500002471.rn.1 | TC0500000401.rn.1 | RTA TC |
| Rat Expression Set 230 | 1373427_PM_at | - | - |
| Rat Expression Set 230 | 1373427_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10868125 | - | - |
| Rat Genome U34 Set | rc_AA851192_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 22227799 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1370703 | - | - |

Rragd
Genomic Map