Assay Details
| Gene Symbol: Mitd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001009714.1 | NP_001009714.1 | 183 | 97 | ||
| XM_006244828.2 | XP_006244890.1 | 257 | 97 | |||
| XM_008767036.1 | XP_008765258.1 | 257 | 97 | |||
| GenBank mRNA | BC088263.1 | - | 183 | 7376620 | 97 | |
More Information
Updated Mapping Information
Set Membership: |
|

Mitd1
Genomic Map