Assay Details
| Gene Symbol: Arhgef4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006244728.2 | XP_006244790.1 | 2 - 3 | 543 | 69 | |
| XM_008766981.1 | XP_008765203.1 | 5 - 6 | 3927 | 69 | ||
More Information
Set Membership: |
|

Arhgef4
Genomic Map