Assay Details
| Gene Symbol: Mfsd9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001108215.1 | NP_001101685.1 | 2 - 3 | 278 | 67 | |
| XM_006244823.2 | XP_006244885.1 | 2 - 3 | 270 | 67 | ||
| XM_006244824.2 | XP_006244886.1 | 2 - 3 | 414 | 67 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Mfsd9
Genomic Map