Assay Details
| Gene Symbol: Mrps9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001100549.1 | NP_001094019.1 | 9 - 10 | 947 | 80 | |
| XM_008767019.1 | XP_008765241.1 | 9 - 10 | 958 | 80 | ||
| GenBank mRNA | BC088242.1 | - | 8 - 9 | 759 | 80 | |
| BC158704.1 | - | 9 - 10 | 915 | 9030006 | 80 | |
More Information
Updated Mapping Information
Set Membership: |
|

Mrps9
Genomic Map