Assay Details
| Gene Symbol: Cryga | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006245063.2 | XP_006245125.1 | 4 - 5 | 815 | 96 | |
| XM_006245064.2 | XP_006245126.1 | 4 - 5 | 592 | 96 | ||
More Information
Set Membership: |
|

Cryga
Genomic Map