Assay Details
| Gene Symbol: Tyw5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001170473.1 | NP_001163944.1 | 3 - 4 | 579 | 73 | |
| NR_033170.1 | 3 - 4 | 560 | 73 | |||
| XM_006244910.2 | XP_006244972.1 | 3 - 4 | 323 | 73 | ||
| GenBank mRNA | BC158619.1 | - | 3 - 4 | 560 | 73 | |
| FM112098.1 | - | 3 - 4 | 294 | 73 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Tyw5
Genomic Map