Assay Details
| Gene Symbol: Fam126b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001025710.1 | NP_001020881.1 | 9 - 10 | 841 | 83 | |
| XM_006244983.2 | XP_006245045.1 | 8 - 9 | 848 | 83 | ||
| XM_006244984.2 | XP_006245046.1 | 8 - 9 | 847 | 83 | ||
| XM_006244985.2 | XP_006245047.1 | 9 - 10 | 881 | 83 | ||
| XM_008767100.1 | XP_008765322.1 | 8 - 9 | 1485 | 83 | ||
| XM_008767101.1 | XP_008765323.1 | 9 - 10 | 1097 | 83 | ||
| XM_008767102.1 | XP_008765324.1 | 9 - 10 | 882 | 83 | ||
| XM_008767103.1 | XP_008765325.1 | 8 - 9 | 1396 | 83 | ||
| XM_008767104.1 | XP_008765326.1 | 8 - 9 | 845 | 83 | ||
| XM_008767105.1 | XP_008765327.1 | 8 - 9 | 848 | 83 | ||
| GenBank mRNA | BC097998.1 | - | 9 - 10 | 841 | 7453086 | 83 |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P127986 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 174941 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 552828 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 647267 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 310973 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 582457 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 847503 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 24310 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 957536 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 840511 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 659362 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 627038 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 953194 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 711859 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 879051 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 449304 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 616893 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 853170 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 653185 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 849526 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 506631 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 123875 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 272566 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 376233 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 654843 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 973612 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 225799 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 701567 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 288947 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 689985 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 431709 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 740425 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 803169 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 130096 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 3373 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 764216 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 177579 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 352350 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 495775 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 94832 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 74973 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 630438 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 347117 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 651943 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 202462 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 181102 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 586705 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 388309 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 958337 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 613720 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 109125 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 415975 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 933691 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 215235 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 427933 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 110112 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 129378 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 113644 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 10160 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 2994 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 170063 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 7502 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 169054 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 121299 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 66908 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P127986 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P127986 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P127986 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 21605 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 91919 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 86171 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 28470 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 78254 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 101742 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10928276 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10928284 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10928288 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10928286 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10928287 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10928283 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10928280 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10928289 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10928285 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10928282 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10928278 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10928281 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10928275 | - | - |
| Rat Genome 230 2.0 Array | 1392614_at | - | - |
| Rat Genome 230 2.0 Array | 1382818_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1397259_at | - | - |
| Rat Genome 230 2.0 Array | 1397259_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1382818_at | - | - |
| Rat Genome 230 2.0 Array | 1388548_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1388548_at | - | - |
| Rat Genome 230 2.0 Array | 1392614_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1388548_at | - | - |
| Affymetrix HT Rat Focus Array | 1397259_at | - | - |
| Affymetrix HT Rat Focus Array | 1397259_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1382818_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1392614_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1382818_at | - | - |
| Affymetrix HT Rat Focus Array | 1388548_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1392614_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR0900016734.rn.1 | TC0900002030.rn.1 | RTA TC |
| Rat Expression Set 230 | 1388548_at | - | - |
| Rat Expression Set 230 | 1388548_PM_at | - | - |
| Rat Expression Set 230 | 1392614_at | - | - |
| Rat Expression Set 230 | 1382818_at | - | - |
| Rat Expression Set 230 | 1392614_PM_at | - | - |
| Rat Expression Set 230 | 1397259_at | - | - |
| Rat Expression Set 230 | 1382818_PM_at | - | - |
| Rat Expression Set 230 | 1397259_PM_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10928275 | - | - |
| Rat Genome U34 Set | rc_AI009875_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 20713565 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1362466 | - | - |

Fam126b
Genomic Map