Assay Details
| Gene Symbol: Ino80d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001191809.1 | NP_001178738.1 | 2 - 3 | 948 | 57 | |
| XM_006245023.2 | XP_006245085.1 | 5 - 6 | 1317 | 57 | ||
| XM_006245024.2 | XP_006245086.1 | 4 - 5 | 2650 | 57 | ||
| XM_006245025.2 | XP_006245087.1 | 4 - 5 | 1213 | 57 | ||
| XM_006245026.2 | XP_006245088.1 | 5 - 6 | 1560 | 57 | ||
| XM_006245029.2 | XP_006245091.1 | 5 - 6 | 1427 | 57 | ||
| XM_006245031.2 | XP_006245093.1 | 4 - 5 | 1323 | 57 | ||
| XM_006245032.2 | XP_006245094.1 | 4 - 5 | 1456 | 57 | ||
| XM_006245033.2 | XP_006245095.1 | 4 - 5 | 1232 | 57 | ||
| XM_006245034.2 | XP_006245096.1 | 4 - 5 | 1122 | 57 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ino80d
Genomic Map