Assay Details
| Gene Symbol: Rab23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001109005.1 | NP_001102475.1 | 3 - 4 | 496 | 71 | |
| XM_008766955.1 | XP_008765177.1 | 3 - 4 | 1157 | 71 | ||
| XM_008766956.1 | XP_008765178.1 | 3 - 4 | 1162 | 71 | ||
| XM_008766957.1 | XP_008765179.1 | 3 - 4 | 482 | 71 | ||
| XM_008766958.1 | XP_008765180.1 | 3 - 4 | 824 | 71 | ||
| XM_008766959.1 | XP_008765181.1 | 3 - 4 | 1155 | 71 | ||
| XM_008766960.1 | XP_008765182.1 | 3 - 4 | 1150 | 71 | ||
| XM_008766961.1 | XP_008765183.1 | 1 - 2 | 1873 | 71 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Rab23
Genomic Map