Assay Details
| Gene Symbol: Fam168b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001271134.1 | NP_001258063.1 | 5 - 6 | 651 | 62 | |
| XM_008766974.1 | XP_008765196.1 | 5 - 6 | 1304 | 62 | ||
| XM_008766975.1 | XP_008765197.1 | 5 - 6 | 1279 | 62 | ||
| XM_008766976.1 | XP_008765198.1 | 4 - 5 | 1266 | 62 | ||
| XM_008766977.1 | XP_008765199.1 | 4 - 5 | 1241 | 62 | ||
| XM_008766978.1 | XP_008765200.1 | 6 - 7 | 752 | 62 | ||
| XM_008766979.1 | XP_008765201.1 | 3 - 4 | 1187 | 62 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Fam168b
Genomic Map