Assay Details
| Gene Symbol: Als2cr11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001024363.1 | NP_001019534.2 | 13 | 1799 | 68 | |
| GenBank mRNA | BC083917.1 | - | 13 | 1799 | 7134752 | 68 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P775974 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 541109 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 261026 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 195692 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 795089 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 589221 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 600434 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 853124 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 743027 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 16681 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 369256 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 72188 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 118445 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 560498 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 791817 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 86199 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 296977 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 356109 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 247428 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 172328 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 780713 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 860928 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 189164 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 707285 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 795968 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 217414 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 564562 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 656989 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 236196 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 96829 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 362183 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 181481 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 163186 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 127075 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 75562 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 735167 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 99611 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 126888 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 52618 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 49766 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 5996 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P775974 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P775974 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P775974 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P775974 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 92824 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 12200 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 86519 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 21619 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 36801 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10928351 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10928347 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10928340 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10928349 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10928341 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10928343 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10928345 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10928350 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10928346 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10928339 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10928348 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10928344 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10928342 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10928338 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10928337 | - | - |
| Rat Genome 230 2.0 Array | 1385766_at | - | - |
| Rat Genome 230 2.0 Array | 1385766_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1385766_at | - | - |
| Affymetrix HT Rat Focus Array | 1385766_PM_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR0900016846.rn.1 | TC0900002037.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0900016848.rn.1 | TC0900002037.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0900016850.rn.1 | TC0900002037.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0900016847.rn.1 | TC0900002037.rn.1 | RTA TC |
| Rat Expression Set 230 | 1385766_PM_at | - | - |
| Rat Expression Set 230 | 1385766_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10928337 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 21255134 | - | - |
| ABI Rat Genome Survey Microarray | 21871979 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1348988 | - | - |

Als2cr11
Genomic Map