Assay Details
| Gene Symbol: Cnnm3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001106901.2 | NP_001100371.2 | 1 - 2 | 1273 | 69 | |
| XM_006244756.2 | XP_006244818.1 | 1 - 2 | 1283 | 69 | ||
| XM_006244757.2 | XP_006244819.1 | 1 - 2 | 1283 | 69 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cnnm3
Genomic Map