Assay Details
| Gene Symbol: Cnnm3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001106901.2 | NP_001100371.2 | 2 - 3 | 1418 | 65 | |
| XM_006244757.2 | XP_006244819.1 | 2 - 3 | 1428 | 65 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cnnm3
Genomic Map