Assay Details
| Gene Symbol: Als2cr11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001024363.1 | NP_001019534.2 | 1 - 2 | 377 | 61 | |
| GenBank mRNA | BC083917.1 | - | 1 - 2 | 377 | 7134752 | 61 |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the BC083917.1 transcripts.
Set Membership: |
|

Als2cr11
Genomic Map