Assay Details
| Gene Symbol: Als2cr11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001024363.1 | NP_001019534.2 | 2 - 3 | 464 | 123 | |
| GenBank mRNA | BC083917.1 | - | 2 - 3 | 464 | 7134752 | 123 |
More Information
Updated Mapping Information
Set Membership: |
|

Als2cr11
Genomic Map