Assay Details
| Gene Symbol: Fcna | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_031348.2 | NP_112638.2 | 7 - 8 | 579 | 61 | |
| XM_008761614.1 | XP_008759836.1 | 7 - 8 | 602 | 61 | ||
| GenBank mRNA | AB026057.2 | - | 7 - 8 | 490 | 61 | |
| BC088128.1 | - | 7 - 8 | 579 | 7376326 | 61 | |
More Information
Updated Mapping Information
Set Membership: |
|

Fcna
Genomic Map