Assay Details
| Gene Symbol: Ptrh1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003749449.3 | XP_003749497.2 | 4 - 5 | 561 | 111 | |
| GenBank mRNA | FQ220792.1 | - | 4 - 5 | 446 | 111 | |
| FQ229591.1 | - | 2 - 3 | 69 | 111 | ||
More Information
Set Membership: |
|

Ptrh1
Genomic Map