Assay Details
| Gene Symbol: RGD1564854 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001109341.1 | NP_001102811.1 | 4 - 5 | 377 | 138 | |
| XM_006233991.2 | XP_006234053.1 | 4 - 5 | 445 | 138 | ||
More Information
Updated Mapping Information
Set Membership: |
|

RGD1564854
Genomic Map