Assay Details
| Gene Symbol: Fam166a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001025694.2 | NP_001020865.1 | 6 - 7 | 785 | 75 | |
| XM_008761589.1 | XP_008759811.1 | 7 - 8 | 768 | 75 | ||
| XM_008761590.1 | XP_008759812.1 | 7 - 8 | 760 | 75 | ||
| XM_008761591.1 | XP_008759813.1 | 6 - 7 | 708 | 75 | ||
| XM_008761592.1 | XP_008759814.1 | 6 - 7 | 584 | 75 | ||
| XM_008761593.1 | XP_008759815.1 | 5 - 6 | 623 | 75 | ||
| XM_008761594.1 | XP_008759816.1 | 4 - 5 | 441 | 75 | ||
| XM_008761595.1 | XP_008759817.1 | 3 - 4 | 387 | 75 | ||
| XM_008761596.1 | XP_008759818.1 | 7 - 8 | 838 | 75 | ||
| GenBank mRNA | BC097396.1 | - | 6 - 7 | 786 | 7453380 | 75 |
More Information
Updated Mapping Information
Set Membership: |
|

Fam166a
Genomic Map