Assay Details
| Gene Symbol: Tor1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_153303.2 | NP_695215.2 | 3 - 4 | 662 | 84 | |
| GenBank mRNA | AY054303.1 | - | 3 - 4 | 644 | 84 | |
| BC078714.1 | - | 3 - 4 | 662 | 7134604 | 84 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tor1a
Genomic Map